6-Maleimidohexanoic acid
6-Maleimidohexanoic acid
6-Maleimidohexanoic acid
6-Maleimidohexanoic acid
Thermo Scientific Chemicals

6-Maleimidohexanoic acid

CAS: 55750-53-3 | C10H13NO4 | 211.217 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 046384.22
also known as 046384-22
Price (USD)
509.65
Special offer
Online exclusive
Ends: 30-Jun-2026
599.00
Save 89.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
509.65
Special offer
Online exclusive
Ends: 30-Jun-2026
599.00
Save 89.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS55750-53-3
IUPAC Name6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanoic acid
Molecular FormulaC10H13NO4
InChI KeyWOJKKJKETHYEAC-UHFFFAOYSA-N
SMILESOC(=O)CCCCCN1C(=O)C=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Weight loss @ 100°C: TGA ≤1.0 wt%
Assay (special)Weight loss @ 200°C: TGA ≤10.0 %
Assay (special)Weight loss @ 300°C: TGA ≤35.0 wt%
Identification (FTIR)Conforms
Appearance (Color)White to pale yellow
View more
6-Maleimidohexanoic acid is used as a probe for introducing maleimides groups into biomolecules and active pharmaceutical ingredients. Further, it is used with N-hydroxysuccinimide ester as a bifunctional cross-linking reagent. In addition to this, it acts as a probe for thiol groups in membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only