Non-wetting Pt 5% Au crucible with RF rim, Top Dia 44mm, BotDia 26.5mm, Ht 49mm, Base Thickness 0.38mm, Capacity 50mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Non-wetting Pt 5% Au crucible with RF rim, Top Dia 44mm, BotDia 26.5mm, Ht 49mm, Base Thickness 0.38mm, Capacity 50mL, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
046389.KT
also known as 046389-KT
1 Ea.
Catalog number 046389.KT
also known as 046389-KT
Price (USD)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Quantity1 Ea.