Titanium gauze, 100 mesh woven from 0.05mm (0.002in) dia wire
Titanium gauze, 100 mesh woven from 0.05mm (0.002in) dia wire
Titanium gauze, 100 mesh woven from 0.05mm (0.002in) dia wire
Thermo Scientific Chemicals

Titanium gauze, 100 mesh woven from 0.05mm (0.002in) dia wire

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
Catalog number 046406.FL
also known as 046406-FL
Price (USD)
136.00
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
136.00
Each
Specifications
CAS7440-32-6
Chemical Name or MaterialTitanum gauze
EINECS Number231-142-3
IUPAC Nametitanium
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only