Polyester methacrylate, PEM-665
Thermo Scientific Chemicals

Polyester methacrylate, PEM-665

Good solubility in both aliphatic and aromatic co-monomers
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 046408.22
also known as 046408-22
Price (USD)
583.65
Online Exclusive
648.00
Save 64.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
583.65
Online Exclusive
648.00
Save 64.35 (10%)
Each
Specifications
Chemical Name or MaterialPolyester methacrylate
ColorYellow
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Good solubility in both aliphatic and aromatic co-monomers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only