Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
Thermo Scientific Chemicals

Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct

CAS: 32993-05-8 | C43H41ClOP2Ru | 772.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 046430.03
also known as 046430-03
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Specifications
CAS32993-05-8
Chemical Name or MaterialChloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
ColorOrange
Formula Weight772.27
Health Hazard 1H302+H312+H332-H315-H319-H335
View more
Cyclopentadienylbis(triphenylphosphine)ruthenium Chloride is used as an efficient transfer hydrogenation catalyst. It is used in the formation of indoles, isoquinolines and quinolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only