Palladium, 1% on granular carbon, reduced
Palladium, 1% on granular carbon, reduced
Palladium, 1% on granular carbon, reduced
Thermo Scientific Chemicals

Palladium, 1% on granular carbon, reduced

CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 046462.14
also known as 046462-14
Price (USD)
117.65
Online Exclusive
130.00
Save 12.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
117.65
Online Exclusive
130.00
Save 12.35 (10%)
Each
Specifications
CAS5-3-7440
ChEBICHEBI:33363
Chemical Name or MaterialPalladium
IUPAC Namepalladium
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
View more
It is used in hydrogenation of aromatic and aliphatic nitro groups; in reductive alkylation/amination; hydrogenation of aromatic carbonyls, olefins, nitriles and as a catalyst in the Suzuki reaction and Stille reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only