Diaion™ HP20, synthetic adsorbent resin, highly porous type
Thermo Scientific Chemicals

Diaion™ HP20, synthetic adsorbent resin, highly porous type

 
Have Questions?
Catalog NumberQuantity
046488.A1
also known as 046488-A1
1 kg
Catalog number 046488.A1
also known as 046488-A1
Price (USD)
188.65
Online Exclusive
209.00
Save 20.35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
188.65
Online Exclusive
209.00
Save 20.35 (10%)
Each
Specifications
Chemical Name or MaterialDiaion™ HP20, synthetic adsorbent resin
Density1.01 g/mL
Name NoteHighly porous type
Particle Size≥250μm
Physical FormBeads
View more
Diaion™ HP20 is used as a filler in chromatography columns due to its extremely large pores. It is considered as an effective adsorbent to eliminate toxic effects in fermentation and separation of traces heavy metal ions. It is also useful for the purification of peptides, proteins, fermentation products, polyphenols and cephalosporin C. It helps to remove bitter tasting substances from fruit juices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only