Polyethylene glycol diacrylate, M.W. 1,000
Polyethylene glycol diacrylate, M.W. 1,000
Polyethylene glycol diacrylate, M.W. 1,000
Thermo Scientific Chemicals

Polyethylene glycol diacrylate, M.W. 1,000

CAS: 26570-48-9 | C5H10O4 | 134.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 046493.03
also known as 046493-03
Price (USD)
95.65
Online exclusive
106.00
Save 10.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
95.65
Online exclusive
106.00
Save 10.35
Each
Chemical Identifiers
CAS26570-48-9
IUPAC Nameethane-1,2-diol; prop-2-enoic acid
Molecular FormulaC5H10O4
InChI KeyFCDZZFLRLVQGEH-UHFFFAOYSA-N
SMILESOCCO.OC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to cream to pale yellow
FormViscous liquid or low melting solid
Assay from Suppliers CofA≥95%
CommentContains inhibitor: <1000ppm 4-methoxyphenol
Molecular WeightMn 1000
PEGDA is used in dental compounds, offset inks, varnishes and flexible coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only