Molybdenum gauze, 10 mesh woven from 0.38mm (0.015mm) dia wire
Molybdenum gauze, 10 mesh woven from 0.38mm (0.015mm) dia wire
Molybdenum gauze, 10 mesh woven from 0.38mm (0.015mm) dia wire
Thermo Scientific Chemicals

Molybdenum gauze, 10 mesh woven from 0.38mm (0.015mm) dia wire

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Catalog number 046494.FL
also known as 046494-FL
Price (USD)
40.70
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
40.70
Each
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
FormGauze, 0.38 mm (0.015 in.) dia. wire
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only