Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd 13.5-15.5%, Thermo Scientific Chemicals
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd 13.5-15.5%, Thermo Scientific Chemicals
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd 13.5-15.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd 13.5-15.5%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
046503.031 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 046503.03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS72287-26-4
IUPAC NameDichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II)
Molecular FormulaC34H28Cl2FeP2Pd
SMILES[Fe].Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)731.71
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pd 13.5-15.5%
Used as a catalyst for Suzuki and Stile couplings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only