Diaion™ UBK550(Na), ion exchange fractionation resin, 8% cross-linked, strongly acidic gel type, 1.9 meq/ml on PS-DVB
Diaion™ UBK550(Na), ion exchange fractionation resin, 8% cross-linked, strongly acidic gel type, 1.9 meq/ml on PS-DVB
Thermo Scientific Chemicals

Diaion™ UBK550(Na), ion exchange fractionation resin, 8% cross-linked, strongly acidic gel type, 1.9 meq/ml on PS-DVB

 
Have Questions?
Catalog NumberQuantity
046609.A1
also known as 046609-A1
1 kg
Catalog number 046609.A1
also known as 046609-A1
Price (USD)
86.65
Online Exclusive
96.10
Save 9.45 (10%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
86.65
Online Exclusive
96.10
Save 9.45 (10%)
Each
Add to cart
Specifications
Chemical Name or MaterialDiaion™ UBK550 (Na), ion exchange fractionation resin
Name Note8% cross-linked, strongly acidic gel type, 1.9meq/mL on PS-DVB
Particle Size220μm
Physical Form220μm Beads
Recommended StorageAmbient temperatures
View more
Chromatographic oligosaccharide sugar separations, sugar alcohol separations

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only