Sodium bis(trifluoromethylsulfonyl)imide
Sodium bis(trifluoromethylsulfonyl)imide
Sodium bis(trifluoromethylsulfonyl)imide
Thermo Scientific Chemicals

Sodium bis(trifluoromethylsulfonyl)imide

CAS: 91742-21-1 | C2HF6NNaO4S2 | 304.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 046640.03
also known as 046640-03
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS91742-21-1
IUPAC Name1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide sodium
Molecular FormulaC2HF6NNaO4S2
InChI KeyQXZNUMVOKMLCEX-UHFFFAOYSA-N
SMILES[Na].FC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (Titration)≥94%
Sodium bis(trifluoromethylsulfonyl)imide is used as an electrolyte in batteries and fuel cells. It also finds use as an ionic liquid and Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only