Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), Pd 12.1%
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), Pd 12.1%
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), Pd 12.1%
Thermo Scientific Chemicals

Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II), Pd 12.1%

CAS: 205319-10-4 | C39H32Cl2OP2Pd | 755.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 046665.03
also known as 046665-03
Price (USD)
170.65
Special offer
Online exclusive
Ends: 28-Dec-2026
201.00
Save 30.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
170.65
Special offer
Online exclusive
Ends: 28-Dec-2026
201.00
Save 30.35
Each
Chemical Identifiers
CAS205319-10-4
IUPAC Name[5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphane; dichloropalladiumbis(ylium)
Molecular FormulaC39H32Cl2OP2Pd
InChI KeyMNBAPNBMBOYNAW-UHFFFAOYSA-L
SMILESCl[Pd++]Cl.CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pd 12.1% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only