Magnesium bis(trifluoromethylsulfonyl)imide
Magnesium bis(trifluoromethylsulfonyl)imide
Magnesium bis(trifluoromethylsulfonyl)imide
Thermo Scientific Chemicals

Magnesium bis(trifluoromethylsulfonyl)imide

CAS: 133395-16-1 | C4F12MgN2O8S4 | 584.576 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 046667.03
also known as 046667-03
Price (USD)
137.65
Online Exclusive
153.00
Save 15.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
137.65
Online Exclusive
153.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS133395-16-1
IUPAC Namemagnesium(2+) bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Molecular FormulaC4F12MgN2O8S4
InChI KeyDMFBPGIDUUNBRU-UHFFFAOYSA-N
SMILES[Mg++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder and/ or granules
Appearance (Color)White to pale yellow
Fluorine-19 NMRConforms
Magnesium bis(trifluoromethylsulfonyl)imide is a strong Lewis acid and used as a catalyst in organic synthesis. It is an electrolyte, which shows high anodic stability used in rechargeable magnesium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only