Hastelloy C∣r gauze, 40 mesh woven from 0.19mm (0.0075in) dia wire
Thermo Scientific Chemicals

Hastelloy C∣r gauze, 40 mesh woven from 0.19mm (0.0075in) dia wire

Have Questions?
Change viewbuttonViewtableView
Quantity:
150 x 150 mm
300 x 300 mm
Catalog number 046690.GT
also known as 046690-GT
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Quantity:
150 x 150 mm
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Specifications
Chemical Name or MaterialHastelloy C™ gauze
MDL NumberMFCD00801065
Mesh Size40 Mesh
Name Note40 mesh woven from 0.19mm (0.0075 in.) dia. wire
Recommended StorageAmbient temperatures
View more
Hastelloy C is used in combustion chambers, heat-treating equipment, collector rings, combustion chambers and exhaust stacks for high-heat applications in for conventional aircraft and jets, pipe connections, pumps and valves for oil refinery, chemical and petrochemical applications. Also used in components exposed to flue gas or flue gas desulfurization plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only