Stainless Steel gauze, 325 mesh woven from 0.036mm (0.0014in) dia wire, Type 316
Thermo Scientific Chemicals

Stainless Steel gauze, 325 mesh woven from 0.036mm (0.0014in) dia wire, Type 316

Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Catalog number 046715.GT
also known as 046715-GT
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.70
Save 7.05 (14%)
Each
Quantity:
150 x 150 mm
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.70
Save 7.05 (14%)
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Chemical Name or MaterialStainless Steel gauze, Type 316
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only