11-Maleimidoundecanoic acid
11-Maleimidoundecanoic acid
11-Maleimidoundecanoic acid
Thermo Scientific Chemicals

11-Maleimidoundecanoic acid

CAS: 57079-01-3 | C15H23NO4 | 281.352 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 046716.14
also known as 046716-14
Price (USD)
180.65
Special offer
Online exclusive
Ends: 30-Jun-2026
212.00
Save 31.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
180.65
Special offer
Online exclusive
Ends: 30-Jun-2026
212.00
Save 31.35 (15%)
Each
Chemical Identifiers
CAS57079-01-3
IUPAC Name11-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)undecanoic acid
Molecular FormulaC15H23NO4
InChI KeyUVZTZBRGZXIBLZ-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCCCCN1C(=O)C=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Appearance (Color)White to off-white
Purity≥95.0%
The maleimide functional group can be used to conjugate a variety of biomolecules such as enzymes and DNA to the polymer chain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only