Tantalum gauze, 80 mesh woven from 0.08mm (0.003 in.) dia. wire, Thermo Scientific Chemicals
Tantalum gauze, 80 mesh woven from 0.08mm (0.003 in.) dia. wire, Thermo Scientific Chemicals
Tantalum gauze, 80 mesh woven from 0.08mm (0.003 in.) dia. wire, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum gauze, 80 mesh woven from 0.08mm (0.003 in.) dia. wire, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
046720.FL
also known as 046720-FL
75 x 75 mm
Catalog number 046720.FL
also known as 046720-FL
Price (USD)
103.00
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
103.00
Each
Specifications
CAS7440-25-7
ChEBICHEBI:33348
Chemical Name or MaterialTantalum gauze
EINECS Number231-13505
IUPAC Nametantalum
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only