Bisphenol F acetate propionate, epoxy curative, EC-298
Bisphenol F acetate propionate, epoxy curative, EC-298
Bisphenol F acetate propionate, epoxy curative, EC-298
Thermo Scientific Chemicals

Bisphenol F acetate propionate, epoxy curative, EC-298

Well suited for use in epoxy/(meth)acrylate hybrid thermoset compositions | CAS: 1071466-53-9 | C18H18O4 | 298.338 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 046724.14
also known as 046724-14
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.30
Save 8.65 (14%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.30
Save 8.65 (14%)
Each
Specifications
CAS1071466-53-9
Chemical Name or MaterialBisphenol F acetate propionate
ColorYellow
Density1.13
Flash Point150°C (302°F)
View more
Bisphenol F acetate propionate does not impede free radical cure, thermoset adhesives, used as curative for epoxy/(meth) acrylate hybrids, hybrid epoxy/free radical thermosets. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only