Polyethylene glycol dimethacrylate, M.W. 3,400
Polyethylene glycol dimethacrylate, M.W. 3,400
Polyethylene glycol dimethacrylate, M.W. 3,400
Thermo Scientific Chemicals

Polyethylene glycol dimethacrylate, M.W. 3,400

CAS: 25852-47-5 | CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 046728.06
also known as 046728-06
Price (USD)
472.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
472.00
Each
Specifications
CAS25852-47-5
Chemical Name or MaterialPolyethylene glycol dimethacrylate
ColorWhite
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Polyethylene glycol dimethacrylate is used as adhesives, coatings, sealants, photo resists, solder masks and photopolymers. Polyethylene Glycol Dimethacrylate forms homopolymers and copolymers. Copolymers of Polyethylene Glycol Dimethacrylate can be prepared with acrylic acid and its salts, amides and esters, and with methacrylates, acrylonitrile, maleic acid esters, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene, unsaturated polyesters and drying oils. It is also used as a very useful feedstock for chemical syntheses. It is employed as a cross linker for PVC plastisols. Polyethylene Glycol 200 Dimethacrylate is incorporated in cosmetic encapsulation resins and dental polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only