Inconel∣r 600 gauze, 40 mesh woven from 0.25mm (0.01in) dia wire
Thermo Scientific Chemicals

Inconel∣r 600 gauze, 40 mesh woven from 0.25mm (0.01in) dia wire

Have Questions?
Change viewbuttonViewtableView
Quantity:
150 x 150 mm
300 x 300 mm
Catalog number 046745.GT
also known as 046745-GT
Price (USD)
41.50
Each
Quantity:
150 x 150 mm
Request bulk or custom format
Price (USD)
41.50
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeNi:Cr:Fe; 77:15.5:7.5 wt%
Chemical Name or MaterialInconel™ 600 gauze
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
Applications include heaters, stills, bubble towers and condensers for processing of fatty acids; evaporator tubes, tube sheets and flaking trays for the manufacture of sodium sulfide; and equipment for handling abietic acid in the manufacture of paper pulp. It is used for retorts, muffles, roller hearths and other furnace components and for heat-treating baskets and trays. The alloy is a standard material of construction for nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only