Palladium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis)
Palladium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis)
Palladium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis)
Thermo Scientific Chemicals

Palladium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis)

CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Catalog NumberQuantity
046783.KG1 Pc.
Catalog number 046783.KG
Price (USD)
-
Quantity:
1 Pc.
Request bulk or custom format
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver white
Assay from Supplier's CofA>99.95% (metals basis)
FormSlug, 6.35mm (0.25in) diameter x 12.7mm (0.50in) length,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only