Palladium, 10% on activated carbon powder, Type A402023-10, nominally 50% water wet
Palladium, 10% on activated carbon powder, Type A402023-10, nominally 50% water wet
Palladium, 10% on activated carbon powder, Type A402023-10, nominally 50% water wet
Thermo Scientific Chemicals

Palladium, 10% on activated carbon powder, Type A402023-10, nominally 50% water wet

CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 046789.04
also known as 046789-04
Price (USD)
-
Quantity:
2 g
Request bulk or custom format
Specifications
CAS5-3-7440
ChEBICHEBI:33363
Chemical Name or MaterialPalladium
ColorBlack
IUPAC Namepalladium
View more
Palladium is used as catalyst. It is also used in jewelry, dentistry, watch making, blood sugar test strips, aircraft spark plugs and in the production of surgical instruments and electrical contacts. It is also used to make professional transverse flutes. The second-biggest application of palladium in electronics is in the manufacture of multilayer ceramic capacitors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only