Inconel∣r 600 gauze, 200 mesh woven from 0.05mm (0.002in) dia wire
Thermo Scientific Chemicals

Inconel∣r 600 gauze, 200 mesh woven from 0.05mm (0.002in) dia wire

Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
Catalog number 046791.FL
also known as 046791-FL
Price (USD)
56.70
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
56.70
Each
Specifications
Chemical Name or MaterialInconel™ 600 gauze
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
MDL NumberMFCD01741927
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only