Polyoxyethylene bis(amine), M.W. 1,000
Polyoxyethylene bis(amine), M.W. 1,000
Polyoxyethylene bis(amine), M.W. 1,000
Thermo Scientific Chemicals

Polyoxyethylene bis(amine), M.W. 1,000

CAS: 24991-53-5 | H2NCH2CH2(OCH2CH2)nNH2
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 046833.06
also known as 046833-06
Price (USD)
559.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
559.00
Each
Chemical Identifiers
CAS24991-53-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Assay by NMR95% min
FormSolid or powder
It is used as PEG modifier, its main products include high-qualified intermediates, various of amino acid and APIs. This Polymer used in the preparation of enzyme conjugates soluble in organic solvents. Promising drug carrier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only