1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-^m-hydrotetracarbonyldiruthenium(II), 98%
Thermo Scientific Chemicals

1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-^m-hydrotetracarbonyldiruthenium(II), 98%

CAS: 104439-77-2 | C62H42O6Ru2
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 046884.02
also known as 046884-02
Price (USD)
143.65
Online Exclusive
159.00
Save 15.35 (10%)
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
143.65
Online Exclusive
159.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS104439-77-2
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min
1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-μ-hydroteracarbonyldiruthenium(II) is widely used in various types of applications in organic synthesis. It is used for hydrogenation of carbonyls, alkenes and imines. It is also used in the alkylation of amines and amination of propargyl alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Calayst for hydrogen transfers including reduction of ketones and aldehydes, and oxidation and/or racemization of amines and alcohols
RUO – Research Use Only