Potassium bis(trifluoromethylsulfonyl)imide
Potassium bis(trifluoromethylsulfonyl)imide
Potassium bis(trifluoromethylsulfonyl)imide
Potassium bis(trifluoromethylsulfonyl)imide
Thermo Scientific Chemicals

Potassium bis(trifluoromethylsulfonyl)imide

CAS: 90076-67-8 | C2F6KNO4S2 | 319.234 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 046909.MD
also known as 046909-MD
Price (USD)
36.10
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
36.10
Each
Chemical Identifiers
CAS90076-67-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Supplier's CofA≥94%

Potassium bis(trifluoromethylsulfonyl)imide is used as Flavor & Fragrance intermediates. It is used as an intermediate in Industries such as agrochemical, dyestuff, pharmaceutical, syntheses material, chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only