Platinum standard form crucible, Top OD 44mm, Bot Dia 26.5mm, Ht 49mm, Base Thickness 0.38mm, Capacity 50mL
Thermo Scientific Chemicals
Platinum standard form crucible, Top OD 44mm, Bot Dia 26.5mm, Ht 49mm, Base Thickness 0.38mm, Capacity 50mL
Have Questions?
Catalog NumberQuantity
046931.KT
also known as 046931-KT
1 Ea.
Catalog number 046931.KT
also known as 046931-KT
Price (USD)
-
Quantity:
1 Ea.
Request bulk or custom format
Specifications
Quantity1 Ea.