Diaion™ PK228(Na), ion exchange resin, 14% cross-linked, strongly acidic porous type, 2.05 meq/ml on PS-DVB
Diaion™ PK228(Na), ion exchange resin, 14% cross-linked, strongly acidic porous type, 2.05 meq/ml on PS-DVB
Diaion™ PK228(Na), ion exchange resin, 14% cross-linked, strongly acidic porous type, 2.05 meq/ml on PS-DVB
Thermo Scientific Chemicals

Diaion™ PK228(Na), ion exchange resin, 14% cross-linked, strongly acidic porous type, 2.05 meq/ml on PS-DVB

CAS: 63182-08-1 | C18H19NaO3S | 338.40 g/mol
Have Questions?
Catalog NumberQuantity
046935.A1
also known as 046935-A1
1 kg
Catalog number 046935.A1
also known as 046935-A1
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS63182-08-1
SpecificationsSpecification SheetSpecification Sheet
Capacity2.05 meq/ml, min
Particle Size95% less than 1180 micron and 1% less than 300 micron
Formbeads
Loss on Drying37 to 43% water
Appearance (Color)Brown
Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation. Used in solutions of high electrolyte content

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation
  • Used in solutions of high electrolyte content
RUO – Research Use Only