Tetrakisepoxy cyclosiloxane
Tetrakisepoxy cyclosiloxane
Tetrakisepoxy cyclosiloxane
Thermo Scientific Chemicals

Tetrakisepoxy cyclosiloxane

CAS: 257284-60-9 | C28H56O12Si4 | 697.084 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 046960.14
also known as 046960-14
Price (USD)
68.65
Online Exclusive
75.80
Save 7.15 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
68.65
Online Exclusive
75.80
Save 7.15 (9%)
Each
Chemical Identifiers
CAS257284-60-9
IUPAC Name2,4,6,8-tetramethyl-2,4,6,8-tetrakis({3-[(oxiran-2-yl)methoxy]propyl})-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Molecular FormulaC28H56O12Si4
InChI KeyBLZDTVROPKIJLZ-UHFFFAOYNA-N
SMILESC[Si]1(CCCOCC2CO2)O[Si](C)(CCCOCC2CO2)O[Si](C)(CCCOCC2CO2)O[Si](C)(CCCOCC2CO2)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofAEpoxy Equivalent Weights (g/eq.): Report
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only