Viscosity standard, Specpure™, nominally 10cSt @ 40 and 2.7cSt @ 100
Thermo Scientific Chemicals

Viscosity standard, Specpure™, nominally 10cSt @ 40 and 2.7cSt @ 100

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Have Questions?
Catalog NumberQuantity
047036.AP
also known as 047036-AP
500 mL
Catalog number 047036.AP
also known as 047036-AP
Price (USD)
124.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
124.00
Each
Chemical Identifiers
CAS8042-47-5
SpecificationsSpecification SheetSpecification Sheet
Viscosity?0.5 cSt at 40?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only