Manganese(III) acetate dihydrate, 97%
Manganese(III) acetate dihydrate, 97%
Manganese(III) acetate dihydrate, 97%
Manganese(III) acetate dihydrate, 97%
Manganese(III) acetate dihydrate, 97%
Thermo Scientific Chemicals

Manganese(III) acetate dihydrate, 97%

CAS: 19513-05-4 | C6H9MnO6 | 232.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 047050.04
also known as 047050-04
Price (USD)
143.65
Online Exclusive
159.00
Save 15.35 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
143.65
Online Exclusive
159.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS19513-05-4
IUPAC Namemanganese(3+) triacetate
Molecular FormulaC6H9MnO6
InChI KeyAHSBSUVHXDIAEY-UHFFFAOYSA-K
SMILES[Mn+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min
Manganese(III) acetate dihydrate, is used as a mild and selective oxidizing agent. It is also a reagent used for radical cyclizations and α-keto-acetoxylation. Mangese triacetate has been used as a single electron oxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only