(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 97%
(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 97%
(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 97%
Thermo Scientific Chemicals

(-)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 97%

CAS: 136705-77-6 | C31H48F3O3P2RhS | 722.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number 047087.ME
also known as 047087-ME
Price (USD)
-
Quantity:
500 mg
Request bulk or custom format
Specifications
CAS136705-77-6
Chemical Name or Material[(R,R)Ethyl-DuPhosRh(COD)]CF3SO3
ColorOrange to Red
Formula Weight722.62
Health Hazard 1Warning
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only