Silicon(IV) oxide, 99.95% (metals basis)
Silicon(IV) oxide, 99.95% (metals basis)
Silicon(IV) oxide, 99.95% (metals basis)
Silicon(IV) oxide, 99.95% (metals basis)
Thermo Scientific Chemicals

Silicon(IV) oxide, 99.95% (metals basis)

CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 047098.22
also known as 047098-22
Price (USD)
181.65
Special offer
Online exclusive
Ends: 30-Jun-2026
213.00
Save 31.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
181.65
Special offer
Online exclusive
Ends: 30-Jun-2026
213.00
Save 31.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Used as sodium stabilizing counterion, for preparation of silica catalytic supports. Also used in glass manufacturing, foundry sand casting, metallurgical flux sand, ceramics sand, gas recovery, paint and coatings, filtration sand, chromatographic column treatment, wood treatment, inks, desiccants and powder metal molding. Its is principle gateway in electronic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only