Manganese(II) fluoride, Puratronic, 99.99% (metals basis)
Manganese(II) fluoride, Puratronic, 99.99% (metals basis)
Manganese(II) fluoride, Puratronic, 99.99% (metals basis)
Thermo Scientific Chemicals

Manganese(II) fluoride, Puratronic, 99.99% (metals basis)

CAS: 7782-64-1 | F2Mn | 92.935 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 047109.14
also known as 047109-14
Price (USD)
296.65
Online exclusive
329.00
Save 32.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
296.65
Online exclusive
329.00
Save 32.35
Each
Chemical Identifiers
CAS7782-64-1
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Manganese(II) fluoride is used in the production of special kinds of glass and lasers. It reacts with antimony pentafluoride in hydrofluoric acid to prepare a complex, Mn(SbF6)2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only