Rhodium, 5% on alumina powder, Type 526, reduced
Rhodium, 5% on alumina powder, Type 526, reduced
Rhodium, 5% on alumina powder, Type 526, reduced
Thermo Scientific Chemicals

Rhodium, 5% on alumina powder, Type 526, reduced

CAS: 7440-16-6 | Rh | 102.91 g/mol
Have Questions?
Catalog number 047120.09
also known as 047120-09
Price (USD)
-
Request bulk or custom format
Specifications
CAS7440-16-6
ChEBICHEBI:33359
Chemical Name or MaterialRhodium
EINECS Number231-125-0
IUPAC Namerhodium
View more

Rhodium, 5% on alumina is used as a catalyst for hydrogenation reactions, nitrous oxide decomposition. Used in manufacturing of nitric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only