Calcium iodide, ultra dry, 99.995% (metals basis)
Calcium iodide, ultra dry, 99.995% (metals basis)
Calcium iodide, ultra dry, 99.995% (metals basis)
Thermo Scientific Chemicals

Calcium iodide, ultra dry, 99.995% (metals basis)

CAS: 10102-68-8 | CaI2 | 293.887 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 047121.14
also known as 047121-14
Price (USD)
170.65
Special offer
Online exclusive
Ends: 30-Jun-2026
201.00
Save 30.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
170.65
Special offer
Online exclusive
Ends: 30-Jun-2026
201.00
Save 30.35 (15%)
Each
Chemical Identifiers
CAS10102-68-8
IUPAC Namecalcium diiodide
Molecular FormulaCaI2
InChI KeyUNMYWSMUMWPJLR-UHFFFAOYSA-L
SMILES[Ca++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Metal Impurities0.005% max.
Particle Size-10 Mesh
FormBead
It is used in photography. It is also used in cat food as a source of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only