Methylcyclopentadienylmanganese tricarbonyl, 97%
Methylcyclopentadienylmanganese tricarbonyl, 97%
Methylcyclopentadienylmanganese tricarbonyl, 97%
Thermo Scientific Chemicals

Methylcyclopentadienylmanganese tricarbonyl, 97%

CAS: 12108-13-3 | C9H7MnO3
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Catalog number 047125.22
also known as 047125-22
Price (USD)
827.65
Online Exclusive
919.00
Save 91.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
827.65
Online Exclusive
919.00
Save 91.35 (10%)
Each
Chemical Identifiers
CAS12108-13-3
IUPAC Name1-methylcyclopenta-2,4-dien-1-ide tris(methanidylidyneoxidanium) manganese
Molecular FormulaC9H7MnO3
InChI KeyIYSGJCJSRBFZSZ-UHFFFAOYSA-N
SMILES[Mn].[C-]#[O+].[C-]#[O+].[C-]#[O+].C[C-]1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange or yellow-orange
FormLiquid
Assay (unspecified)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only