Specifications
Chemical Name or MaterialTellurium, AAS standard solution
DOT InformationTransport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: HYDROCHLORIC ACID
EINECS Number231-595-7
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
View more