Iron(II,III) oxide, nanopowder, 97%
Iron(II,III) oxide, nanopowder, 97%
Iron(II,III) oxide, nanopowder, 97%
Thermo Scientific Chemicals

Iron(II,III) oxide, nanopowder, 97%

CAS: 1317-61-9 | Fe3O4 | 231.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 047141.14
also known as 047141-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1317-61-9
IUPAC Namediiron(3+) λ²-iron(2+) tetraoxidandiide
Molecular FormulaFe3O4
InChI KeyWTFXARWRTYJXII-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Assay (unspecified)96.1% min
Particle Size50-100nm
pH6-8
Surface Area8.5-11.5 m²/g
View more
Iron(II,III) oxide is mainly used as a black-pigment-catalyst in the Haber process and in the water gas shift reaction. It is used as a contrast-agent in MRI scanning and as a pigment in magnetic applications, polishing compounds, cosmetics, medicines, polymers, rubber, fillers, building, construction, appliances and magnetic inks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only