Cobalt, Oil based standard solution, Specpure™ Co 1000μg/g
Cobalt, Oil based standard solution, Specpure™ Co 1000μg/g
Cobalt, Oil based standard solution, Specpure™ Co 1000μg/g
Thermo Scientific Chemicals

Cobalt, Oil based standard solution, Specpure™ Co 1000μg/g

CAS: 7697-37-2 | HNO3 | 63.01 g/mol
Have Questions?
Catalog NumberQuantity
047143.18
also known as 047143-18
50 g
Catalog number 047143.18
also known as 047143-18
Price (USD)
85.65
Online Exclusive
95.40
Save 9.75 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
85.65
Online Exclusive
95.40
Save 9.75 (10%)
Each
Specifications
CAS7697-37-2
ChEBICHEBI:48107
Chemical Name or MaterialCobalt
IUPAC Namenitric acid
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only