Yttrium(III) oxide, nanopowder, 99.995% (REO)
Yttrium(III) oxide, nanopowder, 99.995% (REO)
Yttrium(III) oxide, nanopowder, 99.995% (REO)
Yttrium(III) oxide, nanopowder, 99.995% (REO)
Thermo Scientific Chemicals

Yttrium(III) oxide, nanopowder, 99.995% (REO)

CAS: 1314-36-9 | O3Y2 | 225.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 047154.22
also known as 047154-22
Price (USD)
259.00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
259.00
Each
Add to cart
Chemical Identifiers
CAS1314-36-9
IUPAC Namediyttrium(3+) trioxidandiide
Molecular FormulaO3Y2
InChI KeyRUDFQVOCFDJEEF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Y+3].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.005% max.
Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only