Copper bis(trifluoromethylsulfonyl)imide
Copper bis(trifluoromethylsulfonyl)imide
Copper bis(trifluoromethylsulfonyl)imide
Thermo Scientific Chemicals

Copper bis(trifluoromethylsulfonyl)imide

CAS: 291300-50-0 | C2CuF6NO4S2 | 343.681 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 047162.03
also known as 047162-03
Price (USD)
141.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
141.00
Each
Chemical Identifiers
CAS291300-50-0
IUPAC Nameλ¹-copper(1+) trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Molecular FormulaC2CuF6NO4S2
InChI KeyMSFHCZUIBARZOS-UHFFFAOYSA-N
SMILES[Cu+].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale blue
Assay from Supplier's CofA≥95.0%
FormPowder or crystals or crystalline powder or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only