Tungsten gauze, 150 mesh woven from 0.02mm (0.0008in) dia wire
Tungsten gauze, 150 mesh woven from 0.02mm (0.0008in) dia wire
Tungsten gauze, 150 mesh woven from 0.02mm (0.0008in) dia wire
Thermo Scientific Chemicals

Tungsten gauze, 150 mesh woven from 0.02mm (0.0008in) dia wire

CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
75 x 75 mm
150 x 150 mm
Catalog number 047167.FL
also known as 047167-FL
Price (USD)
96.65
Special offer
Online exclusive
Ends: 28-Dec-2026
113.00
Save 16.35
Each
Quantity:
75 x 75 mm
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 28-Dec-2026
113.00
Save 16.35
Each
Specifications
CAS7440-33-7
ChEBICHEBI:27998
Chemical Name or MaterialTungsten gauze
EINECS Number231-143-9
IUPAC Nametungsten
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only