Aluminum iodide, ultra dry, 99.999% (metals basis)
Aluminum iodide, ultra dry, 99.999% (metals basis)
Aluminum iodide, ultra dry, 99.999% (metals basis)
Thermo Scientific Chemicals

Aluminum iodide, ultra dry, 99.999% (metals basis)

CAS: 7784-23-8 | AlI3 | 407.695 g/mol
Have Questions?
Catalog NumberQuantity
047170.06
also known as 047170-06
5 g
Catalog number 047170.06
also known as 047170-06
Price (USD)
508.65
Online exclusive
565.00
Save 56.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
508.65
Online exclusive
565.00
Save 56.35
Each
Chemical Identifiers
CAS7784-23-8
IUPAC Namealuminium(3+) triiodide
Molecular FormulaAlI3
InChI KeyCECABOMBVQNBEC-UHFFFAOYSA-K
SMILES[Al+3].[I-].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormPowder
Total Metal Impurities0.001% max.
It is employed as a reagent for the scission of certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only