Product Image
Thermo Scientific Chemicals

Yttrium, Oil based standard solution, Specpure™ Y 1000μg/g

 
Have Questions?
Catalog NumberQuantity
047172.18
also known as 047172-18
50 g
Catalog number 047172.18
also known as 047172-18
Price (USD)
86.90
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
86.90
Each
Specifications
Chemical Name or MaterialYttrium, Oil based standard solution
MDL NumberMFCD00011468
Name NoteY 1000μg/mL
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only