Chromium(II) chloride, 99.9% (metals basis)
Chromium(II) chloride, 99.9% (metals basis)
Chromium(II) chloride, 99.9% (metals basis)
Thermo Scientific Chemicals

Chromium(II) chloride, 99.9% (metals basis)

CAS: 10049-05-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 047176.03
also known as 047176-03
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.10
Save 6.45
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.10
Save 6.45
Each
Chemical Identifiers
CAS10049-05-5
IUPAC Nameλ²-chromium(2+) dichloride
Molecular FormulaCl2Cr
InChI KeyXBWRJSSJWDOUSJ-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Chromium(II) chloride is used in the condensation of aldehydes with trisubstituted chloroalkenes as well as in the synthesis of a highly active precatalyst for ethylene oligimerization. Further uses of chromium(II) chloride include the conversion of terminal olefins into trans-iodocyclopropanes (in conjunction with iodoform and tetraethyl ethylenediamine) with excellent results; generating bis(indenyl)chromium dimers and other various related products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only