Ruthenium(III) chloride, anhydrous, Ru 47.7% min
Ruthenium(III) chloride, anhydrous, Ru 47.7% min
Ruthenium(III) chloride, anhydrous, Ru 47.7% min
Thermo Scientific Chemicals

Ruthenium(III) chloride, anhydrous, Ru 47.7% min

CAS: 10049-08-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 047182.04
also known as 047182-04
Price (USD)
122.00
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
122.00
Each
Chemical Identifiers
CAS10049-08-8
IUPAC Nameruthenium(3+) trichloride
Molecular FormulaCl3Ru
InChI KeyYBCAZPLXEGKKFM-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Ru+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormPowder
Assay (special)≥47.7% (Ruthenium)
Ruthenium(III) chloride is widely used as a starting material of ruthenium complexes. It acts as a catalyst used in the oxidative cyclization of 1,7-dienes to oxepane diols. It is used in the hydroxylation of tertiary hydrocarbons in combination of periodate or bromate. It is involved as a catalyst in the synthesis of 2,3-pyridinedicarboxylic acid-13C3, 15N, where the unlabelled analog is an inhibitor of glucose synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only