Benzyltrimethylammonium hydroxide, 40% w/w aq. soln.
Benzyltrimethylammonium hydroxide, 40% w/w aq. soln.
Benzyltrimethylammonium hydroxide, 40% w/w aq. soln.
Thermo Scientific Chemicals

Benzyltrimethylammonium hydroxide, 40% w/w aq. soln.

CAS: 100-85-6 | C10H17NO | 167.252 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 047189.AE
also known as 047189-AE
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.20
Save 7.55 (14%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.20
Save 7.55 (14%)
Each
Chemical Identifiers
CAS100-85-6
IUPAC Namebenzyltrimethylazanium hydroxide
Molecular FormulaC10H17NO
InChI KeyNDKBVBUGCNGSJJ-UHFFFAOYSA-M
SMILES[OH-].C[N+](C)(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay from Suppliers CofA≥38.0 to ≤42.0%
Benzyltrimethylammonium hydroxide is used as a phase-transfer catalyst. It is also used in aldol condensation reactions and base-catalyzed dehydration reactions. Further, it serves as a strong organic base and used in Horner-Wadsworth-Emmons olefination reactions. In addition, it is an active component in the formulation, which is used in silicon wafer cleaning applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only