Lutetium(III) bromide, ultra dry, 99.99% (metals basis)
Lutetium(III) bromide, ultra dry, 99.99% (metals basis)
Lutetium(III) bromide, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Lutetium(III) bromide, ultra dry, 99.99% (metals basis)

CAS: 14456-53-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 047190.03
also known as 047190-03
Price (USD)
96.65
Online Exclusive
107.00
Save 10.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
96.65
Online Exclusive
107.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS14456-53-2
IUPAC Namelutetium(3+) tribromide
Molecular FormulaBr3Lu
InChI KeyDWHGOINJUKABSY-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Lu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only